Learn More
Electron Microscopy Sciences NIGHTSEA™ Stereo Fluorescence Adapter PULSE lamp base, Royal Blue/Green Only Barrier Filter
SDP
Catalog No. 501929141
Encompass_Preferred
The NIGHTSEA™ Stereo Microscope Fluorescence Adapter adapts just about any stereo microscope (dissecting microscope) for fluorescence with no modification to the microscope itself. The modular design lets you easily switch between several different excitation/emission combinations to work with a variety of fluorescent proteins and other fluorophores. There are six different excitation/emission combinations available, plus white light.Excellent for quick screening of your fluorescent genotypes, genotype sorting, fluorescence-aided dissection, injection, or micromanipulation, freeing up your research-grade fluorescence microscopes for more demanding work, and bringing fluorescence into the teaching laboratory. System consists of flexible gooseneck lamp base with power supply, light head, ring adapter for microscope, barrier filter, and filter shield.Color: Royal Blue / Green OnlyExcitation: 440-460nmEmission filter: 500-560 nm bandpassFluorophores: GFP, eGFP, fluorescein
Catalog No. 50-192-9141 Supplier Electron Microscopy Sciences Supplier No. SFARBGOPULSE
The NIGHTSEA™ Stereo Microscope Fluorescence Adapter adapts just about any stereo microscope (dissecting microscope) for fluorescence with no modification to the microscope itself. The modular design lets you easily switch between several different excitation/emission combinations to work with a variety of fluorescent proteins and other fluorophores. There are six different excitation/emission combinations available, plus white light.Excellent for quick screening of your fluorescent genotypes, genotype sorting, fluorescence-aided dissection, injection, or micromanipulation, freeing up your research-grade fluorescence microscopes for more demanding work, and bringing fluorescence into the teaching laboratory. System consists of flexible gooseneck lamp base with power supply, light head, ring adapter for microscope, barrier filter, and filter shield.Color: Royal Blue / Green OnlyExcitation: 440-460nmEmission filter: 500-560 nm bandpassFluorophores: GFP, eGFP, fluorescein