Learn More
Electron Microscopy Sciences NIGHTSEA™ White Light Head
SDP
Catalog No. 501902699
Encompass_Preferred
A white light head is also available as an illumination option for the Stereo Microscope Fluorescence Adapter. Now if you are using the NIGHTSEA system for fluorescence you do not need a separate light source for your white-light work-just exchange the fluorescence excitation light head module for the white-light module in a matter of seconds. Extremely convenient for general illumination and as a focusing aid for fluorescence imaging.
Catalog No. 50-190-2699 Supplier Electron Microscopy Sciences Supplier No. SFALHWH
A white light head is also available as an illumination option for the Stereo Microscope Fluorescence Adapter. Now if you are using the NIGHTSEA system for fluorescence you do not need a separate light source for your white-light work-just exchange the fluorescence excitation light head module for the white-light module in a matter of seconds. Extremely convenient for general illumination and as a focusing aid for fluorescence imaging.