Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences NIGHTSEA™ Fluorescence Adapter Shield, Green
SDP

Catalog No. 501902705
Encompass_Preferred

Additional combinations excitation/ emission wavelength combination can be added NIGHTSEA™ Stereo Microscope Fluorescence Viewing system by purchasing individual light head, barrier filter, and viewing shield combinations. These three elements can be removed and replaced in seconds, and color coding ensures that you are using the right combination. The barrier filter clicks on to the ring adapter magnetically, so it is easy to remove it to switch back to white light viewing.
Color: Green
Excitation: 510-540nm
Emission filter: 600 nm longpass
Fluorophores: DsRed, dTomato

Catalog No. 50-190-2705 Supplier Electron Microscopy Sciences Supplier No. SFASHGR
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Additional combinations excitation/ emission wavelength combination can be added NIGHTSEA™ Stereo Microscope Fluorescence Viewing system by purchasing individual light head, barrier filter, and viewing shield combinations. These three elements can be removed and replaced in seconds, and color coding ensures that you are using the right combination. The barrier filter clicks on to the ring adapter magnetically, so it is easy to remove it to switch back to white light viewing.
Color: Green
Excitation: 510-540nm
Emission filter: 600 nm longpass
Fluorophores: DsRed, dTomato

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.