Learn More
Electron Microscopy Sciences Nigrosin 25 GR
SDP
Catalog No. 5027485
Encompass_Preferred
(Acid Black 2; Gray R, B, BB; Indulin Black; Nigrosin W, WL ; Silver Gray; Steel Gray)
Solubility: 10.0% Water; 0.0% Alcohol; 1.0% Cellosolve; 11.0% Glycol; 0.0% Xylene
Nigrosin is also known as Acid Black 2; Gray R, B, BB, Indulin Black, Nigrosin W, WL , Silver Gray and Steel Gray. It is used for negative staining of bacteria. With Basic Fuchsin, staining bacteria spores. Dorner, Le Lait, 6, 8 (1926). Biochem. Biophys. Acta 69,159 (1963).
Catalog No. 50-274-85 Supplier Electron Microscopy Sciences Supplier No. 19010
(Acid Black 2; Gray R, B, BB; Indulin Black; Nigrosin W, WL ; Silver Gray; Steel Gray)
Solubility: 10.0% Water; 0.0% Alcohol; 1.0% Cellosolve; 11.0% Glycol; 0.0% Xylene
Nigrosin is also known as Acid Black 2; Gray R, B, BB, Indulin Black, Nigrosin W, WL , Silver Gray and Steel Gray. It is used for negative staining of bacteria. With Basic Fuchsin, staining bacteria spores. Dorner, Le Lait, 6, 8 (1926). Biochem. Biophys. Acta 69,159 (1963).