Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Nigrosin Saturated Aqueous Solution
SCI_ED

Catalog No. S25447 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S25447 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25447

missing translation for 'validMainframeMsg'

This product is an Educational Grade Chemical — no C of A is available.

Specifications

Chemical Name or Material Nigrosin Saturated Aqueous Solution
CAS 7697-37-2, 7732-18-5
Assay Percent Range 0.03
Synonym Nigrosine Biological Stain
Packaging Solution
Density 1g/mL
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.