Learn More
Nigrosin Stain Solution, 100 mL, G-Biosciences
SCI_ED
Catalog No. S001149
Change view
Click to view available options
Quantity:
100 mL
Catalog No. Quantity
S001149 100 mL
1 options
Catalog No. S001149 Supplier G-Biosciences Supplier No. 7862247
Add to Cart
Add to Cart
Nigrosin Stain Solution is used as a negative staining method that permits the visualization of transparent and unstainable capsule of many organisms.
  • Nigrosin stain solution is used for negative staining in microscopy, creating a dark background to highlight unstained structures like bacterial capsules, cell morphology, or the viability of sperm
Specifications
Quantity 100 mL
Product Type Nigrosin Stain Solution