Learn More
Medchemexpress LLC (+)-Niguldipine hydrochloride | 113145-69-0 | MFCD00873564 | 98.1% | 646.17 g/mol | C36H40ClN3O6 | 1 MG
SDP
Catalog No. 5000213185
Encompass_Preferred
(+)-Niguldipine hydrochloride is the S-enantiomer of niguldipine, a dihydropyridine calcium channel antagonist used in research to probe L-type calcium channel-mediated signaling and vasodilation mechanisms. It is supplied as an off-white to yellow solid with high purity for laboratory applications.
  • High purity suitable for analytical and biological assays.
  • S-enantiomer useful for enantiospecific pharmacology studies.
  • Small vials appropriate for screening and mechanistic experiments.
  • Off-white to yellow solid form simplifies handling and formulation.
  • Storage recommendations provided for sealed samples and solvent solutions.
Catalog No. 50-002-13185 Supplier Medchemexpress LLC Supplier No. HY1013901MG
(+)-Niguldipine hydrochloride is the S-enantiomer of niguldipine, a dihydropyridine calcium channel antagonist used in research to probe L-type calcium channel-mediated signaling and vasodilation mechanisms. It is supplied as an off-white to yellow solid with high purity for laboratory applications.
  • High purity suitable for analytical and biological assays.
  • S-enantiomer useful for enantiospecific pharmacology studies.
  • Small vials appropriate for screening and mechanistic experiments.
  • Off-white to yellow solid form simplifies handling and formulation.
  • Storage recommendations provided for sealed samples and solvent solutions.