Learn More
Medchemexpress LLC (+)-Niguldipine hydrochloride | 113145-69-0 | MFCD00873564 | 98.1% | 646.17 g/mol | C36H40ClN3O6 | 5 MG
SDP
Catalog No. 5000213200
Encompass_Preferred
(+)-Niguldipine hydrochloride is the S-enantiomer of niguldipine, a dihydropyridine calcium-channel blocker and α1-adrenergic receptor antagonist used in pharmacological and cardiovascular research. The compound is supplied as a high-purity solid for in vitro and biochemical studies. Certificate of Analysis, data sheet, and SDS are available; handle and store sealed, away from moisture, following the supplier COA.
Catalog No. 50-002-13200 Supplier Medchemexpress LLC Supplier No. HY1013905MG
(+)-Niguldipine hydrochloride is the S-enantiomer of niguldipine, a dihydropyridine calcium-channel blocker and α1-adrenergic receptor antagonist used in pharmacological and cardiovascular research. The compound is supplied as a high-purity solid for in vitro and biochemical studies. Certificate of Analysis, data sheet, and SDS are available; handle and store sealed, away from moisture, following the supplier COA.