Learn More
Cayman Chemical NIguldIpInhydrochlorIde 10mg
SDP
Catalog No. 502984065
Encompass_Preferred
A dihydropyridine that acts as a potent, selective α1A-adrenoceptor antagonist (Ki = 0.16 nM); less potently blocks L- and T-type Ca2+ channels (IC50s = 0.4 and 0.9 µM, respectively)
Catalog No. 50-298-4065 Supplier Cayman Chemical Supplier No. 1953410 MG
A dihydropyridine that acts as a potent, selective α1A-adrenoceptor antagonist (Ki = 0.16 nM); less potently blocks L- and T-type Ca2+ channels (IC50s = 0.4 and 0.9 µM, respectively)