Learn More
Selleck Chemical LLC NIH-12848
SDP
Catalog No. 5000002915
Encompass_Preferred
NIH-12848 is a specific PI5P4K inhibitor with an IC50 of approximately 1 M but does not inhibit the and PI5P4K isoforms at concentrations up to 100 M
Catalog No. 50-000-02915 Supplier Selleck Chemical LLC Supplier No. S65295MG
NIH-12848 is a specific PI5P4K inhibitor with an IC50 of approximately 1 M but does not inhibit the and PI5P4K isoforms at concentrations up to 100 M