Learn More
Medchemexpress LLC HY-101423 5mg Medchemexpress, NIH-12848 CAS:959551-10-1 Purity:>98%
SDP
Catalog No. 501932122
Encompass_Preferred
Medchemexpress, HY-101423 5mg NIH-12848 CAS:959551-10-1 NIH-12848 is a putative phosphatidylinositol 5-phosphate 4-kinase γ ( PI5P4Kγ ) inhibitor with an IC 50 of 1 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2122 Supplier Medchemexpress LLC Supplier No. HY1014235MG
Medchemexpress, HY-101423 5mg NIH-12848 CAS:959551-10-1 NIH-12848 is a putative phosphatidylinositol 5-phosphate 4-kinase γ ( PI5P4Kγ ) inhibitor with an IC 50 of 1 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.