Learn More
Sigma Aldrich Fine Chemicals Biosciences Mouse (Mus musculus) NIH 3T3 Embryonic Fibroblast Cell Line | Contact-Insensitive Proliferation | 1 Vial
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 930615241VL
Encompass
NIH 3T3 Cell Line | Species: Mouse (Mus musculus) | Tissue Source: Embryonic | Morphology: Spindle-shaped fibroblast | Growth Properties: Adherent, contact-insensitive, continuous proliferation | Applications: Cell cycle studies, DNA damage response, primary cilia research, transfection host, stem cell culture, cell-based assays | Keywords: Mouse fibroblast cells, embryonic mouse fibroblasts, contact-insensitive cell proliferation, DNA damage response studies, transfection host cells, cell-based biological assays
Catalog No. NC1741396