Learn More
ATCC Mus musculus
Supplier:  ATCC CRL1658/DEL
Encompass
NIH/3T3 is a fibroblast cell line that was isolated from a mouse NIH/Swiss embryo. This cell line is highly sensitive to sarcoma virus focus formation and leukemia virus propagation and has proven to be very useful in DNA transfection studies.
Catalog No. 50-188-474FP