Learn More
Cell Biolabs Inc NIH3T3/GFP Cell Line
Supplier:  Cell Biolabs Inc AKR214
Encompass_Preferred
Cell Biolabs' NIH3T3/GFP Cell Line stably expresses GFP and blasticidin-resistant genes. Both GFP and blasticidin-resistant genes were introduced into parental NIH3T3 cells using lentivirus. This cell line otherwise exhibits the same characteristics of the parental NIH3T3 cell line.
Catalog No. 50-672-593