Learn More
ATCC NIH:OVCAR-3; Ovarian Adenocarcinoma; Human (Homo sapiens)
Catalog No. 502383854
Encompass
NIH:OVCAR-3 [OVCAR3] are epithelial cells that were isolated in 1982 from the malignant ascites of a patient with progressive adenocarcinoma of the ovary. Use these cells to study drug resistance in ovarian cancer.
Catalog No. 50-238-3854 Supplier ATCC Supplier No. HTB161
NIH:OVCAR-3 [OVCAR3] are epithelial cells that were isolated in 1982 from the malignant ascites of a patient with progressive adenocarcinoma of the ovary. Use these cells to study drug resistance in ovarian cancer.