Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NIK-IN-2 | 1202763-91-4 | C20H22N4O3 | 5 MG
SDP

Catalog No. 5000373743
Encompass_Preferred

NIK-IN-2 is a potent inhibitor of NF-κB-inducing kinase (NIK) with a pIC50 of 7.4. It is an alkynylpyrimidine-based covalent inhibitor that targets a unique cysteine in NF-κB-inducing kinase, playing an important role in cancer research.

  • Potent inhibitor of NF-κB-inducing kinase (NIK)
  • pIC50 of 7.4
  • Alkynylpyrimidine-based covalent inhibitor
  • Appears as a white to light yellow solid
  • Purity of 99.00%
  • Soluble in DMSO (33.33 mg/mL)
  • Inhibits NIK in human HT-29 cells with an IC50 of 0.02 μM

Catalog No. 50-003-73743 Supplier Medchemexpress LLC Supplier No. HY1535165MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NIK-IN-2 is a potent inhibitor of NF-κB-inducing kinase (NIK) with a pIC50 of 7.4. It is an alkynylpyrimidine-based covalent inhibitor that targets a unique cysteine in NF-κB-inducing kinase, playing an important role in cancer research.

  • Potent inhibitor of NF-κB-inducing kinase (NIK)
  • pIC50 of 7.4
  • Alkynylpyrimidine-based covalent inhibitor
  • Appears as a white to light yellow solid
  • Purity of 99.00%
  • Soluble in DMSO (33.33 mg/mL)
  • Inhibits NIK in human HT-29 cells with an IC50 of 0.02 μM

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.