Learn More
Novus Biologicals™ NIK/MAP3K14 Antibody Blocking Peptide

Specifications
Specifications
| Gene ID (Entrez) | 9020 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| For Use With (Application) | This peptide is useful as a blocking peptide for NBP2-23603. For further blocking peptide related protocol, click here . |
| Gene Alias | EC 2.7.11.25, FTDCR1B, HS, HsNIK, HSNIKserine/threonine protein-kinase, mitogen-activated protein kinase kinase kinase 14, NIKNF-kappa-beta-inducing kinase, Serine/threonine-protein kinase NIK |
| Gene Symbol | MAP3K14 |
| Quantity | 0.1 mg |
| Research Category | Apoptosis, Cancer, Protein Kinase, Signal Transduction |
| Storage Buffer | Peptide dissolved in dH20. Contains no BSA. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.