Learn More
Selleck Chemical LLC NIK SMI1 5mg
SDP
Supplier:  Selleck Chemical LLC S89415MG
Encompass_Preferred
NIK SMI1 is a highly potent and selective NF-?B-inducing kinase (NIK) inhibitor with Ki of 0.23 nM for NIK-catalyzed hydrolysis of ATP to ADP. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3448