Learn More
Medchemexpress LLC Nikkomycin Z | 59456-70-1 | 98.6% | 495.44 | 1 MG
SDP
Catalog No. 5000401497
Encompass_Preferred
Nikkomycin Z is an orally active nucleoside peptide antifungal agent. It functions by inhibiting chitin synthesis, acting as a competitive analogue of the chitin synthase substrate UDP-N-acetylglucosamine. This compound demonstrates antifungal activity and enhances efficacy when combined with echinocandins such as Caspofungin and Micafungin. In vivo studies show it reduces fungal burden and extends survival in murine models of *Histoplasma capsulatum* infection.
  • Inhibits chitin synthesis
  • Competitive analogue of UDP-N-acetylglucosamine
  • Active against *Candida albicans* and *Candida parapsilosis*
  • Enhances efficacy with echinocandins
  • Reduces fungal burden in *Histoplasma capsulatum* infection models
Catalog No. 50-004-01497 Supplier Medchemexpress LLC Supplier No. HY195931MG
Nikkomycin Z is an orally active nucleoside peptide antifungal agent. It functions by inhibiting chitin synthesis, acting as a competitive analogue of the chitin synthase substrate UDP-N-acetylglucosamine. This compound demonstrates antifungal activity and enhances efficacy when combined with echinocandins such as Caspofungin and Micafungin. In vivo studies show it reduces fungal burden and extends survival in murine models of *Histoplasma capsulatum* infection.
  • Inhibits chitin synthesis
  • Competitive analogue of UDP-N-acetylglucosamine
  • Active against *Candida albicans* and *Candida parapsilosis*
  • Enhances efficacy with echinocandins
  • Reduces fungal burden in *Histoplasma capsulatum* infection models