Learn More
Evoqua Water Technologies LLC NIL-8 Residual Test Strips, W2T934634, 50 test strips per container
Catalog No. NC3237189
Encompass
NIL-8 Residual Test Strips are easy to use and conveniently packaged 50 test strips per container. Each strip has a "reaction zone" pad located at one end of the strip. When stored and used properly, the test strip pad can indicate residual NIL-8.
Catalog No. NC3237189 Supplier Evoqua Water Technologies LLC Supplier No. W2T934634
NIL-8 Residual Test Strips are easy to use and conveniently packaged 50 test strips per container. Each strip has a "reaction zone" pad located at one end of the strip. When stored and used properly, the test strip pad can indicate residual NIL-8.