Learn More
Evoqua Water Technologies LLC NIL-8 Test Strips, W2T934633 50 test strips
Catalog No. NC3237184
Encompass
NIL-8 Working Concentration Test Strips are easy to use and conveniently packaged 50 test strips per container. Each strip has a "reaction zone" pad located at one end of the strip. When stored and used properly, the test strip pad can indicate the working concentration of NIL-8.
Catalog No. NC3237184 Supplier Evoqua Water Technologies LLC Supplier No. W2T934633
NIL-8 Working Concentration Test Strips are easy to use and conveniently packaged 50 test strips per container. Each strip has a "reaction zone" pad located at one end of the strip. When stored and used properly, the test strip pad can indicate the working concentration of NIL-8.