Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Nile Blue A 10 GR
SDP

Catalog No. 5027486
Encompass_Preferred

(Basic Blue 12, Nile Blue Sulfate)
CAS #3625-57-8 (C₂₀H₂₀N₃O)₂SO₄ F.W. 732.84
Solubility: 6.0% Water; 5.0% Alcohol; 5.0% Cellosolve; 5.0% Glycol; 0.25% Xylene

Differentiates melamins and lipofuscins; Used in fat staining. Lillie, Stain Tech., 31,151 (1956); J. Histochem., 4,377; Z. Anal. Chem. 260, 290 (1972). For staining bone sections. Cytologia, 11, 261 (1940)

Catalog No. 50-274-86 Supplier Electron Microscopy Sciences Supplier No. 19020
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Basic Blue 12, Nile Blue Sulfate)
CAS #3625-57-8 (C₂₀H₂₀N₃O)₂SO₄ F.W. 732.84
Solubility: 6.0% Water; 5.0% Alcohol; 5.0% Cellosolve; 5.0% Glycol; 0.25% Xylene

Differentiates melamins and lipofuscins; Used in fat staining. Lillie, Stain Tech., 31,151 (1956); J. Histochem., 4,377; Z. Anal. Chem. 260, 290 (1972). For staining bone sections. Cytologia, 11, 261 (1940)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.