Learn More
Electron Microscopy Sciences Nile Blue A, Staining Solution 500 ML
SDP
Catalog No. 5031846
Encompass_Preferred
A fat and lipid stain used for the differentiation of melamines and lipofuchsins and staining for phospholipids.
Catalog No. 50-318-46 Supplier Electron Microscopy Sciences Supplier No. 2651101
A fat and lipid stain used for the differentiation of melamines and lipofuchsins and staining for phospholipids.