Learn More
Polysciences, Inc. Nile Blue Acrylamide, 2089099-00-1, MFCD31382389, 100MG
SDP
Catalog No. NC1060749
Encompass_Preferred
Nile Blue Acrylamide, Formula: C23H22N3O2CI, Molecular Weight: 408, Appearance: Dark blue crystals, Storage: Protect from light. Store at -20C. Nile Blue dye itself is a fluorescent dye that is used routinely in histology to impart a blue color to cell nuclei, where it highlights the distinction between neutral lipids (triglycerides, etc.) which are stained pink and fatty acids, which are stained blue.
Catalog No. NC1060749 Supplier Polysciences, Inc. Supplier No. 25395100
Nile Blue Acrylamide, Formula: C23H22N3O2CI, Molecular Weight: 408, Appearance: Dark blue crystals, Storage: Protect from light. Store at -20C. Nile Blue dye itself is a fluorescent dye that is used routinely in histology to impart a blue color to cell nuclei, where it highlights the distinction between neutral lipids (triglycerides, etc.) which are stained pink and fatty acids, which are stained blue.