Learn More
Medchemexpress LLC Nile blue chloride 1g
SDP
Catalog No. 5000204984
Encompass_Preferred
Nile blue chloride is a highly fluorescent and photostable organic dye Nile blue chloride and fluorescein isothiocyanate (FITC) can be used to construct a ratiometric pH sensitive probe for tracking the pH of the extracellular fluid between cancer cells in realtime Nile Blue chloride has the potential for the research of nonlinear optics[1][2]
Catalog No. 50-002-04984 Supplier Medchemexpress LLC Supplier No. HYW1108981G
Nile blue chloride is a highly fluorescent and photostable organic dye Nile blue chloride and fluorescein isothiocyanate (FITC) can be used to construct a ratiometric pH sensitive probe for tracking the pH of the extracellular fluid between cancer cells in realtime Nile Blue chloride has the potential for the research of nonlinear optics[1][2]