Learn More
Medchemexpress LLC HY-D0718 100mg , Nile Red CAS:7385-67-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYD0718100MG
Encompass_Preferred
HY-D0718 100mg Nile Red CAS: 7385-67-3 Nile Red (Nile Blue A oxazone) is a selective and hydrophobic fluorescent stain for intracellular lipid droplets and neutral lipids. Nile Red is intensely fluorescent in all organic solvents and the fluorescence colors range from golden yellow to deep red. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1158