Learn More
Medchemexpress LLC HY-D0718 10mg Medchemexpress, Nile Red CAS:7385-67-3 Purity:>98%
SDP
Catalog No. 501933175
Encompass_Preferred
Medchemexpress, HY-D0718 10mg Nile Red CAS:7385-67-3 Nile Red is a strongly fluorescent stain in the presence of a hydrophobic environment for the detection of intracellular lipid droplets. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3175 Supplier Medchemexpress LLC Supplier No. HYD071810MG
Medchemexpress, HY-D0718 10mg Nile Red CAS:7385-67-3 Nile Red is a strongly fluorescent stain in the presence of a hydrophobic environment for the detection of intracellular lipid droplets. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.