Learn More
AdipoGen Nile Red, 7385-67-3, MFCD00011639, 1 g
SDP
Catalog No. 501016761
Encompass_Preferred
MF: C20H18N2O2, MW: 318.4, ≥98%, Appearance: Green to brown powder. Uncharged, hydrophobic, photostable fluorescent probe that strongly fluoresces bright red in hydrophobic (lipid-rich) environments, but is almost nonfluorescent in water. This lipophilic stain is commonly used for the detection of intracellular lipid droplets in cells such as adipocytes by fluorescence microscopy and flow cytometry.
Catalog No. 50-101-6761 Supplier AdipoGen Supplier No. CDXN0107G001
MF: C20H18N2O2, MW: 318.4, ≥98%, Appearance: Green to brown powder. Uncharged, hydrophobic, photostable fluorescent probe that strongly fluoresces bright red in hydrophobic (lipid-rich) environments, but is almost nonfluorescent in water. This lipophilic stain is commonly used for the detection of intracellular lipid droplets in cells such as adipocytes by fluorescence microscopy and flow cytometry.