Learn More
Selleck Chemical LLC Nile Red
SDP
Catalog No. 5000002845
Encompass_Preferred
Nile Red (Nile Blue A oxazone Phenoxazone 9) is an excellent vital fluorescent stain for the detection of intracellular lipid droplets in the presence of a hydrophobic environment Nile Red is applied for staining intracellular lipids hydrophobic domains of proteins and lysosomal phospholipid inclusions
Catalog No. 50-000-02845 Supplier Selleck Chemical LLC Supplier No. S681825MG
Nile Red (Nile Blue A oxazone Phenoxazone 9) is an excellent vital fluorescent stain for the detection of intracellular lipid droplets in the presence of a hydrophobic environment Nile Red is applied for staining intracellular lipids hydrophobic domains of proteins and lysosomal phospholipid inclusions