Learn More
Selleck Chemical LLC Nile Red
Supplier: Selleck Chemical LLC S68181G
Nile Red (Nile Blue A oxazone Phenoxazone 9) is an excellent vital fluorescent stain for the detection of intracellular lipid droplets in the presence of a hydrophobic environment Nile Red is applied for staining intracellular lipids hydrophobic domains of proteins and lysosomal phospholipid inclusions