Learn More
Apexbio Technology LLC Nile Red 7385-67-3 50mg
SDP
Catalog No. 5000064070
Encompass_Preferred
Nile Red is a lipophilic fluorescent compound commonly utilized in biological and biomedical research to visualize intracellular lipid droplets Structurally this dye is hydrophobic and selectively partitions into neutral lipid environments within cells enabling targeted staining of lipid-rich regions Upon binding to intracellular lipids Nile Red exhibits enhanced fluorescence emission which can be detected by fluorescent microscopy or flow cytometry Researchers employ Nile Red for studying lipid distribution lipid metabolism and lipid storage dynamics in various cell types Additionally Nile Red is employed within assays exploring metabolic alterations lipid-accumulation disorders and cellular lipid content measurement facilitating analysis of lipid-associated physiological and pathological processes
Catalog No. 50-000-64070 Supplier Apexbio Technology LLC Supplier No. B820950
Nile Red is a lipophilic fluorescent compound commonly utilized in biological and biomedical research to visualize intracellular lipid droplets Structurally this dye is hydrophobic and selectively partitions into neutral lipid environments within cells enabling targeted staining of lipid-rich regions Upon binding to intracellular lipids Nile Red exhibits enhanced fluorescence emission which can be detected by fluorescent microscopy or flow cytometry Researchers employ Nile Red for studying lipid distribution lipid metabolism and lipid storage dynamics in various cell types Additionally Nile Red is employed within assays exploring metabolic alterations lipid-accumulation disorders and cellular lipid content measurement facilitating analysis of lipid-associated physiological and pathological processes