Learn More
Medchemexpress LLC Nile Red | 7385-67-3 | 63.67 UG
SDP
Catalog No. 5000410401
Encompass_Preferred
Nile Red (solution) is a lipophilic stain with environment-sensitive fluorescence, appearing intensely fluorescent in lipid-rich environments and minimally so in aqueous media. It is an excellent vital stain for detecting intracellular lipid droplets via fluorescence microscopy and flow cytofluorometry.
  • Stains intracellular lipid droplets red.
  • Fluorescence wavelength is 559/635 nm.
  • Suitable for in vitro cell staining.
  • Applicable for in vivo studies, such as in Caenorhabditis elegans, to observe lipid bodies.
Catalog No. 50-004-10401 Supplier Medchemexpress LLC Supplier No. HYD0718Y63.67UG
Nile Red (solution) is a lipophilic stain with environment-sensitive fluorescence, appearing intensely fluorescent in lipid-rich environments and minimally so in aqueous media. It is an excellent vital stain for detecting intracellular lipid droplets via fluorescence microscopy and flow cytofluorometry.
  • Stains intracellular lipid droplets red.
  • Fluorescence wavelength is 559/635 nm.
  • Suitable for in vitro cell staining.
  • Applicable for in vivo studies, such as in Caenorhabditis elegans, to observe lipid bodies.