Learn More
Abcam Nile Red, lipophilic stain, 25MG
Catalog No. 502652252
Encompass_Preferred
MW 318.4 Da, Purity >95%. Photostable, lipophilic stain that strongly fluoresces in hydrophobic (lipid-rich) environments, but has minimal fluorescence in aqueous media. Commonly used for the detection of intracellular lipid droplets by fluorescence microscopy and flow cytofluorometry.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-2252 Supplier Abcam Supplier No. AB21940325MG
MW 318.4 Da, Purity >95%. Photostable, lipophilic stain that strongly fluoresces in hydrophobic (lipid-rich) environments, but has minimal fluorescence in aqueous media. Commonly used for the detection of intracellular lipid droplets by fluorescence microscopy and flow cytofluorometry.
The product is subject to the following: Abcam Restricted Use Statement