Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nile red, MP Biomedicals™

Catalog No. ICN15174410 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15174410 10 mg
ICN15174483 250 mg
ICN15174490 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15174410 Supplier MP Biomedicals, Inc Supplier No. 0215174410
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7385-67-3
Molecular Formula C20H18N2O2
Molecular Weight (g/mol) 318.38
MDL Number MFCD00011639
InChI Key VOFUROIFQGPCGE-UHFFFAOYSA-N
Synonym Nile Blue A Oxazone, 9-Diethylamino-5H-benzo[a]phenoxazine-5-one
PubChem CID 65182
ChEBI CHEBI:52169
IUPAC Name 8-(diethylamino)-12H-10-oxa-5-azatetraphen-12-one
SMILES CCN(CC)C1=CC=C2N=C3C(OC2=C1)=CC(=O)C1=CC=CC=C31

Specifications

Color Green
Quantity 10 mg
Physical Form Powder
Formula Weight 318.4
Chemical Name or Material Nile red, Determination of lipids, staining whole tissue
EINECSNumber 230-966-0
Recommended Storage Store at Room Temperature (15° to 30°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.