Learn More
Medchemexpress LLC Nile Red (Nile Blue A oxazone) | 7385-67-3 | 99.7% | 500 MG
SDP
Catalog No. 5000410909
Encompass_Preferred
Nile red (Nile blue oxazone) is a lipophilic stain with environment-sensitive fluorescence, exhibiting intense fluorescence in a lipid-rich environment and minimal fluorescence in aqueous media. It is an excellent vital stain for detecting intracellular lipid droplets via fluorescence microscopy and flow cytometry, staining them red with a fluorescence wavelength of 559/635 nm.
  • Lipophilic stain
  • Environment-sensitive fluorescence
  • Intensely fluorescent in lipid-rich environments
  • Minimal fluorescence in aqueous media
  • Excellent vital stain for detecting intracellular lipid droplets
  • Used with fluorescence microscopy and flow cytometry
  • Stains intracellular lipid droplets red
  • Fluorescence wavelength: 559/635 nm
Catalog No. 50-004-10909 Supplier Medchemexpress LLC Supplier No. HYD0718500MG
Nile red (Nile blue oxazone) is a lipophilic stain with environment-sensitive fluorescence, exhibiting intense fluorescence in a lipid-rich environment and minimal fluorescence in aqueous media. It is an excellent vital stain for detecting intracellular lipid droplets via fluorescence microscopy and flow cytometry, staining them red with a fluorescence wavelength of 559/635 nm.
  • Lipophilic stain
  • Environment-sensitive fluorescence
  • Intensely fluorescent in lipid-rich environments
  • Minimal fluorescence in aqueous media
  • Excellent vital stain for detecting intracellular lipid droplets
  • Used with fluorescence microscopy and flow cytometry
  • Stains intracellular lipid droplets red
  • Fluorescence wavelength: 559/635 nm