Learn More
Medchemexpress LLC Nile Red (Nile blue oxazone) | 7385-67-3 | 99.7% | 5 MG
SDP
Catalog No. 5000410860
Encompass_Preferred
Nile red (Nile blue oxazone) is a lipophilic stain that exhibits environment-sensitive fluorescence. It is intensely fluorescent in lipid-rich environments and shows minimal fluorescence in aqueous media. This stain is excellent for detecting intracellular lipid droplets by fluorescence microscopy and flow cytometry, staining them red. The fluorescence wavelength is 559/635 nm.
  • Lipophilic stain
  • Environment-sensitive fluorescence
  • Intensely fluorescent in lipid-rich environments
  • Minimal fluorescence in aqueous media
  • Excellent vital stain for detecting intracellular lipid droplets
  • Used in fluorescence microscopy and flow cytometry
  • Stains intracellular lipid droplets red
  • Fluorescence wavelength: 559/635 nm
Catalog No. 50-004-10860 Supplier Medchemexpress LLC Supplier No. HYD07185MG
Nile red (Nile blue oxazone) is a lipophilic stain that exhibits environment-sensitive fluorescence. It is intensely fluorescent in lipid-rich environments and shows minimal fluorescence in aqueous media. This stain is excellent for detecting intracellular lipid droplets by fluorescence microscopy and flow cytometry, staining them red. The fluorescence wavelength is 559/635 nm.
  • Lipophilic stain
  • Environment-sensitive fluorescence
  • Intensely fluorescent in lipid-rich environments
  • Minimal fluorescence in aqueous media
  • Excellent vital stain for detecting intracellular lipid droplets
  • Used in fluorescence microscopy and flow cytometry
  • Stains intracellular lipid droplets red
  • Fluorescence wavelength: 559/635 nm