Learn More
Medchemexpress LLC Nile Red (solution) | 7385-67-3 | 31.83 UG
SDP
Catalog No. 5000410491
Encompass_Preferred
Nile Red is a lipophilic stain that exhibits environment-sensitive fluorescence. It is intensely fluorescent in lipid-rich environments and has minimal fluorescence in aqueous media. Nile Red serves as an excellent vital stain for the detection of intracellular lipid droplets by fluorescence microscopy and flow cytometry, staining them red.
  • Environment-sensitive fluorescence
  • Intensely fluorescent in a lipid-rich environment
  • Minimal fluorescence in aqueous media
  • Excellent vital stain for the detection of intracellular lipid droplets
  • Stains intracellular lipid droplets red
  • Fluorescence wavelength is 559/635 nm
Catalog No. 50-004-10491 Supplier Medchemexpress LLC Supplier No. HYD0718Y31.83UG
Nile Red is a lipophilic stain that exhibits environment-sensitive fluorescence. It is intensely fluorescent in lipid-rich environments and has minimal fluorescence in aqueous media. Nile Red serves as an excellent vital stain for the detection of intracellular lipid droplets by fluorescence microscopy and flow cytometry, staining them red.
  • Environment-sensitive fluorescence
  • Intensely fluorescent in a lipid-rich environment
  • Minimal fluorescence in aqueous media
  • Excellent vital stain for the detection of intracellular lipid droplets
  • Stains intracellular lipid droplets red
  • Fluorescence wavelength is 559/635 nm