Learn More
Medchemexpress LLC Nile Red (solution) | 7385-67-3 | 318.37 UG
SDP
Catalog No. 5000409621
Encompass_Preferred
Nile Red solution is a lipophilic stain with environment-sensitive fluorescence, exhibiting intense fluorescence in lipid-rich environments and minimal fluorescence in aqueous media. It serves as an excellent vital stain for detecting intracellular lipid droplets via fluorescence microscopy and flow cytometry.
  • Functions as a lipophilic stain
  • Features environment-sensitive fluorescence
  • Intensely fluorescent in lipid-rich environments
  • Minimally fluorescent in aqueous media
  • Detects intracellular lipid droplets
  • Suitable for fluorescence microscopy and flow cytometry
  • Stains intracellular lipid droplets red
  • Provides a fluorescence wavelength of 559/635 nm
Catalog No. 50-004-09621 Supplier Medchemexpress LLC Supplier No. HYD0718Y318.37UG
Nile Red solution is a lipophilic stain with environment-sensitive fluorescence, exhibiting intense fluorescence in lipid-rich environments and minimal fluorescence in aqueous media. It serves as an excellent vital stain for detecting intracellular lipid droplets via fluorescence microscopy and flow cytometry.
  • Functions as a lipophilic stain
  • Features environment-sensitive fluorescence
  • Intensely fluorescent in lipid-rich environments
  • Minimally fluorescent in aqueous media
  • Detects intracellular lipid droplets
  • Suitable for fluorescence microscopy and flow cytometry
  • Stains intracellular lipid droplets red
  • Provides a fluorescence wavelength of 559/635 nm