Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nile Red, Technical Grade, Sigma-Aldrich

Catalog No. 01259069 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-259-069 1 g
01-259-040 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-259-069 Supplier MilliporeSigma Supelco Supplier No. N30131G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nile Red is lipophilic in nature and exploits the hydrophobic properties of molecules such as lipids and plastics that fluoresce when excited with certain wavelengths, facilitating quantification and identification.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7385-67-3
Color Brown to Green
Quantity 1 g
Molecular Weight (g/mol) 318.37 g/mol
Grade Technical
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.