Learn More
Selleck Chemical LLC Nilotinib S1033-100mg
SDP
Catalog No. 503108499
Encompass_Preferred
Nilotinib is a selective Bcr-Abl inhibitor with IC50 less than 30 nM in Murine myeloid progenitor cells Nilotinib induces autophagy through AMPK activition
Catalog No. 50-310-8499 Supplier Selleck Chemical LLC Supplier No. S1033100MG
Nilotinib is a selective Bcr-Abl inhibitor with IC50 less than 30 nM in Murine myeloid progenitor cells Nilotinib induces autophagy through AMPK activition