Learn More
Selleck Chemical LLC Nilotinib (AMN-107) 25mg 641571-10-0
SDP
Catalog No. NC0394691
Encompass_Preferred
Nilotinib (AMN-107) is a selective Bcr-Abl inhibitor with IC50 less than 30 nM in Murine myeloid progenitor cells. Nilotinib induces autophagy through AMPK activition. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0394691 Supplier Selleck Chemical LLC Supplier No. S103325MG
Nilotinib (AMN-107) is a selective Bcr-Abl inhibitor with IC50 less than 30 nM in Murine myeloid progenitor cells. Nilotinib induces autophagy through AMPK activition. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.