Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nilotinib-d6 | 1268356-17-7 | MFCD15071309 | 100.0% | 535.55 | C28H16D6F3N7O | 10 MG
SDP

Catalog No. 5000247775
Encompass_Preferred

Nilotinib-d6 is a deuterium-labeled analytical standard of nilotinib used for quantitative bioanalytical and mass-spectrometry applications. It is intended as an internal standard for pharmacokinetic, metabolic, and stability studies involving nilotinib, providing consistent isotopic labeling for accurate measurement.

  • Deuterium-labeled internal standard suitable for LC-MS/MS quantitation.
  • High purity for reliable analytical performance.
  • Molecular formula C28H16D6F3N7O and molecular weight 535.55.
  • Supplied in small mass packs appropriate for laboratory use.
  • Useful for pharmacokinetic, metabolic, and stability studies.

Catalog No. 50-002-47775 Supplier Medchemexpress LLC Supplier No. HY10159S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nilotinib-d6 is a deuterium-labeled analytical standard of nilotinib used for quantitative bioanalytical and mass-spectrometry applications. It is intended as an internal standard for pharmacokinetic, metabolic, and stability studies involving nilotinib, providing consistent isotopic labeling for accurate measurement.

  • Deuterium-labeled internal standard suitable for LC-MS/MS quantitation.
  • High purity for reliable analytical performance.
  • Molecular formula C28H16D6F3N7O and molecular weight 535.55.
  • Supplied in small mass packs appropriate for laboratory use.
  • Useful for pharmacokinetic, metabolic, and stability studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.