Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nilotinib-d6 | 1268356-17-7 | MFCD15071309 | 100.0% | 535.55 g/mol | C28H16D6F3N7O | 1 MG
SDP

Catalog No. 5000245468
Encompass_Preferred

Nilotinib-d6 is a deuterium-labeled analog of Nilotinib supplied for research and analytical applications. It serves as a characterized reference standard and internal standard for mass spectrometry and pharmacokinetic studies involving the Bcr-Abl tyrosine kinase inhibitor class. The compound is provided as a high-purity solid with guidance for storage and solubility.

  • Deuterium-labeled analog for use as a research standard.
  • High purity (reported 99.95%).
  • Molecular weight approximately 535.55 g/mol.
  • Molecular formula C28H16D6F3N7O.
  • Available in small milligram quantities suitable for analytical use.
  • Soluble in DMSO with recommended warming and sonication for dissolution.
  • Store powder at -20°C; store solutions at -80°C for long-term stability.

Catalog No. 50-002-45468 Supplier Medchemexpress LLC Supplier No. HY10159S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nilotinib-d6 is a deuterium-labeled analog of Nilotinib supplied for research and analytical applications. It serves as a characterized reference standard and internal standard for mass spectrometry and pharmacokinetic studies involving the Bcr-Abl tyrosine kinase inhibitor class. The compound is provided as a high-purity solid with guidance for storage and solubility.

  • Deuterium-labeled analog for use as a research standard.
  • High purity (reported 99.95%).
  • Molecular weight approximately 535.55 g/mol.
  • Molecular formula C28H16D6F3N7O.
  • Available in small milligram quantities suitable for analytical use.
  • Soluble in DMSO with recommended warming and sonication for dissolution.
  • Store powder at -20°C; store solutions at -80°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.