Learn More
Medchemexpress LLC Nilotinib-d6 | 1268356-17-7 | MFCD15071309 | 99.9% | 535.55 g/mol | C28H16D6F3N7O | 5 MG
SDP
Catalog No. 5000248961
Encompass_Preferred
Nilotinib-d6 is a deuterium-labeled analog of nilotinib supplied as an isotopically labeled reference standard for research and analytical applications. It retains the Bcr-Abl tyrosine kinase inhibitor scaffold and is intended for method development, quantitative mass spectrometry, and stability studies. Not for human or clinical use.
  • Deuterium-labeled reference standard for analytical and tracer studies.
  • Suitable for mass spectrometry and quantitative method development.
  • High isotopic enrichment and chemical purity for reliable results.
  • Supplied in a small, accurately weighed 5 mg pack for precise dosing.
  • Soluble in common organic solvents such as DMSO for easy handling.
Catalog No. 50-002-48961 Supplier Medchemexpress LLC Supplier No. HY10159S5MG
Nilotinib-d6 is a deuterium-labeled analog of nilotinib supplied as an isotopically labeled reference standard for research and analytical applications. It retains the Bcr-Abl tyrosine kinase inhibitor scaffold and is intended for method development, quantitative mass spectrometry, and stability studies. Not for human or clinical use.
  • Deuterium-labeled reference standard for analytical and tracer studies.
  • Suitable for mass spectrometry and quantitative method development.
  • High isotopic enrichment and chemical purity for reliable results.
  • Supplied in a small, accurately weighed 5 mg pack for precise dosing.
  • Soluble in common organic solvents such as DMSO for easy handling.