Learn More
Selleck Chemical LLC Nilotinib hydrochloride S5205-25mg
SDP
Catalog No. 501948244
Encompass_Preferred
Nilotinib hydrochloride (AMN-107) is the hydrochloride salt form of nilotinib an orally bioavailable Bcr-Abl tyrosine kinase inhibitor with antineoplastic activity
Catalog No. 50-194-8244 Supplier Selleck Chemical LLC Supplier No. S520525MG
Nilotinib hydrochloride (AMN-107) is the hydrochloride salt form of nilotinib an orally bioavailable Bcr-Abl tyrosine kinase inhibitor with antineoplastic activity