Learn More
Selleck Chemical LLC Nilotinib hydrochloride S5205-1g
SDP
Catalog No. 503112349
Encompass_Preferred
Nilotinib hydrochloride (AMN-107) is the hydrochloride salt form of nilotinib an orally bioavailable Bcr-Abl tyrosine kinase inhibitor with antineoplastic activity
Catalog No. 50-311-2349 Supplier Selleck Chemical LLC Supplier No. S52051G
Nilotinib hydrochloride (AMN-107) is the hydrochloride salt form of nilotinib an orally bioavailable Bcr-Abl tyrosine kinase inhibitor with antineoplastic activity