Learn More
Medchemexpress LLC Nilotinib hydrochloride dihydrate | 00-00-0 | 99.9% | 602.01 g/mol | C28H27ClF3N7O3 | 200 MG
SDP
Catalog No. 5000248960
Encompass_Preferred
Nilotinib hydrochloride dihydrate is the dihydrate salt of nilotinib, a second-generation Bcr-Abl tyrosine kinase inhibitor used in oncology research. Supplied for research and analytical applications, it is intended for laboratory studies of chronic myelogenous leukemia mechanisms and kinase signaling. Not for human therapeutic use.
  • Potent Bcr-Abl tyrosine kinase inhibitor.
  • Suitable for in vitro and in vivo research models.
  • High purity suitable for analytical applications (99.89%).
  • Supplied as a solid with molecular weight 602.01 g/mol.
  • Available in laboratory pack sizes for experimental use.
Catalog No. 50-002-48960 Supplier Medchemexpress LLC Supplier No. HY10159C200MG
Nilotinib hydrochloride dihydrate is the dihydrate salt of nilotinib, a second-generation Bcr-Abl tyrosine kinase inhibitor used in oncology research. Supplied for research and analytical applications, it is intended for laboratory studies of chronic myelogenous leukemia mechanisms and kinase signaling. Not for human therapeutic use.
  • Potent Bcr-Abl tyrosine kinase inhibitor.
  • Suitable for in vitro and in vivo research models.
  • High purity suitable for analytical applications (99.89%).
  • Supplied as a solid with molecular weight 602.01 g/mol.
  • Available in laboratory pack sizes for experimental use.