Learn More
Medchemexpress LLC Nilotinib hydrochloride dihydrate | 00-00-0 | 99.9% | 602.01 g/mol | C28H27ClF3N7O3 | 25 MG
SDP
Catalog No. 5000248068
Encompass_Preferred
Nilotinib hydrochloride dihydrate is the dihydrate hydrochloride salt of nilotinib, a potent, orally active Bcr-Abl tyrosine kinase inhibitor used in research to study chronic myelogenous leukemia (CML) and Bcr-Abl-driven signaling. It is supplied as a solid research reagent with high purity and documented solubility and storage recommendations to support assay preparation and long-term stability.
  • High purity suitable for analytical and biological studies.
  • Potent Bcr-Abl tyrosine kinase inhibitor for CML research.
  • Solid form with recommended sealed storage away from moisture.
  • Soluble in DMSO at 100 mg/mL for assay preparation.
  • Documented stability in solution and long-term storage conditions.
Catalog No. 50-002-48068 Supplier Medchemexpress LLC Supplier No. HY10159C25MG
Nilotinib hydrochloride dihydrate is the dihydrate hydrochloride salt of nilotinib, a potent, orally active Bcr-Abl tyrosine kinase inhibitor used in research to study chronic myelogenous leukemia (CML) and Bcr-Abl-driven signaling. It is supplied as a solid research reagent with high purity and documented solubility and storage recommendations to support assay preparation and long-term stability.
  • High purity suitable for analytical and biological studies.
  • Potent Bcr-Abl tyrosine kinase inhibitor for CML research.
  • Solid form with recommended sealed storage away from moisture.
  • Soluble in DMSO at 100 mg/mL for assay preparation.
  • Documented stability in solution and long-term storage conditions.