Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nilotinib hydrochloride dihydrate | 00-00-0 | 99.9% | 602.01 g/mol | C28H27ClF3N7O3 | 500 MG
SDP

Catalog No. 5000249256
Encompass_Preferred

Nilotinib hydrochloride dihydrate is the hydrochloride dihydrate salt of nilotinib (AMN107), an orally active Bcr-Abl tyrosine kinase inhibitor used in research on chronic myelogenous leukemia. Supplied as a solid powder for laboratory research, the material is provided with assay data and manufacturer storage guidance for short- and long-term handling.

  • High purity (99.9%).
  • Solid powder form suitable for research use.
  • Molecular formula C28H27ClF3N7O3; molecular weight 602.01 g/mol.
  • Available in multiple pack sizes, including 500 mg.
  • Store sealed at 4°C; in solution store at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-49256 Supplier Medchemexpress LLC Supplier No. HY10159C500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nilotinib hydrochloride dihydrate is the hydrochloride dihydrate salt of nilotinib (AMN107), an orally active Bcr-Abl tyrosine kinase inhibitor used in research on chronic myelogenous leukemia. Supplied as a solid powder for laboratory research, the material is provided with assay data and manufacturer storage guidance for short- and long-term handling.

  • High purity (99.9%).
  • Solid powder form suitable for research use.
  • Molecular formula C28H27ClF3N7O3; molecular weight 602.01 g/mol.
  • Available in multiple pack sizes, including 500 mg.
  • Store sealed at 4°C; in solution store at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.