Learn More
Medchemexpress LLC Nilotinib hydrochloride dihydrate | 00-00-0 | MFCD09833716 | 99.9% | 602.01 g/mol | C28H27ClF3N7O3 | 100 MG
SDP
Catalog No. 5000249040
Encompass_Preferred
Nilotinib hydrochloride dihydrate is the hydrochloride salt, dihydrate form of nilotinib, supplied as a white to off-white solid for research use. It functions as a Bcr-Abl tyrosine kinase inhibitor and is intended for biochemical and cell-based studies of Bcr-Abl signaling and leukemia models, provided in laboratory-scale quantities for in vitro and preclinical experiments.
  • High purity suitable for research-grade assays.
  • Hydrochloride dihydrate salt form for improved handling and reproducibility.
  • Soluble in DMSO at high concentrations (≈100 mg/mL) with ultrasonic assistance.
  • Stable when stored sealed at 4°C; long-term storage in solvent at -80°C.
  • White to off-white powder form for easy weighing and dissolution.
Catalog No. 50-002-49040 Supplier Medchemexpress LLC Supplier No. HY10159C100MG
Nilotinib hydrochloride dihydrate is the hydrochloride salt, dihydrate form of nilotinib, supplied as a white to off-white solid for research use. It functions as a Bcr-Abl tyrosine kinase inhibitor and is intended for biochemical and cell-based studies of Bcr-Abl signaling and leukemia models, provided in laboratory-scale quantities for in vitro and preclinical experiments.
  • High purity suitable for research-grade assays.
  • Hydrochloride dihydrate salt form for improved handling and reproducibility.
  • Soluble in DMSO at high concentrations (≈100 mg/mL) with ultrasonic assistance.
  • Stable when stored sealed at 4°C; long-term storage in solvent at -80°C.
  • White to off-white powder form for easy weighing and dissolution.